SY3H3D67A41E Synthonix Corporation

4-Chloro-2-vinylpyridine

  • MDL number MFCD15475064
  • Cas Number : 98420-89-4
  • Molecular Weight : 139.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to orange to brown liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45320.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45320.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart