SY3H3D67A421 Synthonix Corporation

3-Chloro-2-nitrotoluene

  • MDL number MFCD04117977
  • Cas Number : 5367-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow Solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45350.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45350.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart