SY3H3D67A42F Synthonix Corporation

2-(4-Chloropyridin-2-yl)acetic acid hydrochloride

  • Cas Number : 1688656-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45371.html

Safety Information

MSDS https://synthonix.com/45371.html
Availability details for
Quantity
  • Availability
  • Add to Cart