SY3H3D67A44F Synthonix Corporation

2-Chloro-5-fluoroanisole

  • MDL number MFCD00042573
  • Cas Number : 450-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45592.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45592.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart