SY3H3D67A454 Synthonix Corporation

(4-Chloro-3-fluorophenyl)methanol

  • MDL number MFCD00143292
  • Cas Number : 202925-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45597.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45597.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart