SY3H3D67A476 Synthonix Corporation

4-Chloro-2-iodoanisole

  • MDL number MFCD00052858
  • Cas Number : 52807-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powde - Liquid / Colour: Colorless - White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45602.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45602.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart