SY3H3D67A47A Synthonix Corporation

4-Chlorobenzene-1-carboximidamide hydrochloride

  • MDL number MFCD00126401
  • Cas Number : 14401-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45622.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45622.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart