SY3H3D67A47C Synthonix Corporation

3-Chloro-2-methyl-6-nitroaniline

  • Cas Number : 51123-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45647.html

Safety Information

MSDS https://synthonix.com/45647.html
Availability details for
Quantity
  • Availability
  • Add to Cart