SY3H3D67A485 Synthonix Corporation

4-Chloro-2-methoxy-6-nitroaniline

  • MDL number MFCD28405560
  • Cas Number : 859877-49-9
  • Molecular Weight : 202.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Orange to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45676.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45676.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart