SY3H3D67A497 Synthonix Corporation

3-Chloro-5-fluoro-2-methylaniline

  • Cas Number : 886761-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45801.html

Safety Information

MSDS https://synthonix.com/45801.html
Availability details for
Quantity
  • Availability
  • Add to Cart