SY3H3D67A49D Synthonix Corporation

2-Cyano-3-cyclopropylacrylic acid

  • Cas Number : 98895-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45821.html

Safety Information

MSDS https://synthonix.com/45821.html
Availability details for
Quantity
  • Availability
  • Add to Cart