SY3H3D67A4A9 Synthonix Corporation

3-Chloro-2-methylphenol

  • MDL number MFCD00079738
  • Cas Number : 3260-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to Yellow to Pale-brown or Gray Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45954.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45954.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart