SY3H3D67A4B0 Synthonix Corporation

N-Carbamimidoylisonicotinamide

  • MDL number MFCD18431657
  • Cas Number : 6531-74-4
  • Molecular Weight : 164.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46069.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46069.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart