SY3H3D67A4BC Synthonix Corporation

2-Chloro-4,6-dimethylpyridine

  • MDL number MFCD08277279
  • Cas Number : 30838-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46101.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46101.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart