SY3H3D67A4E2 Synthonix Corporation

3-(Chloromethyl)-5-methylpyridine hydrochloride

  • MDL number MFCD11656303
  • Cas Number : 1007089-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46109.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46109.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart