SY3H3D67A4F2 Synthonix Corporation

4-Chloro-3-methoxyaniline

  • MDL number MFCD00672967
  • Cas Number : 13726-14-2
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Greyish yellow green - Brown - Black
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46190.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46190.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart