SY3H3D67A4F3 Synthonix Corporation

1-(2-Chloropyridin-3-yl)ethanol

  • MDL number MFCD16037360
  • Cas Number : 131674-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46191.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46191.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart