SY3H3D67A4F4 Synthonix Corporation

1-(4-Chloropyridin-2-yl)ethanol

  • MDL number MFCD18802690
  • Cas Number : 121638-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46192.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46192.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart