SY3H3D67A520 Synthonix Corporation

(R)-2-((5-Chloro-4-iodopyridin-2-yl)amino)propan-1-ol

  • MDL number MFCD25459280
  • Cas Number : 1799412-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46707.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46707.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart