SY3H3D67A55D Synthonix Corporation

1-(5-(((2-Cyanoethyl)thio)methyl)thiazol-2-yl)guanidine

  • Cas Number : 131184-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46888.html

Safety Information

MSDS https://synthonix.com/46888.html
Availability details for
Quantity
  • Availability
  • Add to Cart