SY3H3D67A560 Synthonix Corporation

cis-Cyclohexane-1,4-dicarboxylic acid

  • MDL number MFCD00064950
  • Cas Number : 619-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47032.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47032.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart