SY3H3D67A564 Synthonix Corporation

2-Cyclohexylacetonitrile

  • MDL number MFCD00058667
  • Cas Number : 4435-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Colorless to Yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47050.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47050.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart