SY3H3D67A565 Synthonix Corporation

1-Cyclopropylpiperidin-4-one

  • MDL number No MDL found
  • Cas Number : 62813-01-8
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropyl-4-piperidinone
Inchi Code 1S/C8H13NO/c10-8-3-5-9(6-4-8)7-1-2-7/h7H,1-6H2
Inchi Key DTUJRJIWGWTNFQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47058.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47058.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart