SY3H3D67A569 Synthonix Corporation

N-Cyclopropyl-N-methylpyrrolidin-3-amine

  • MDL number MFCD16709553
  • Cas Number : 250275-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47146.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47146.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart