SY3H3D67A56C Synthonix Corporation

(1R,2R)-Cyclohexane-1,2-diyldimethanol

  • MDL number MFCD16660860
  • Cas Number : 65376-05-8
  • Molecular Weight : 144.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47157.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47157.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart