SY3H3D67A57D Synthonix Corporation

4-Chloro-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD00060671
  • Cas Number : 320-41-2
  • Molecular Weight : 205.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-(trifluoromethyl)benzonitrile
Inchi Code 1S/C8H3ClF3N/c9-6-2-1-5(4-13)7(3-6)8(10,11)12/h1-3H
Inchi Key GRNQHTXPUDZMGB-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Refrigerator
Physical Form Light yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47276.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47276.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart