SY3H3D67A58C Synthonix Corporation

2-Cyano-3-nitrobenzoic acid

  • Cas Number : 1260834-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47356.html

Safety Information

MSDS https://synthonix.com/47356.html
Availability details for
Quantity
  • Availability
  • Add to Cart