SY3H3D67A58D Synthonix Corporation

2-Chloro-6-fluoroquinazolin-4(3H)-one

  • MDL number MFCD11520685
  • Cas Number : 769158-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47361.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47361.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart