SY3H3D67A5CD Synthonix Corporation

2-Chloro-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD00792415
  • Cas Number : 39226-97-6
  • Molecular Weight : 224.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47468.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47468.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart