SY3H3D67A5E8 Synthonix Corporation

7-Chloro-1H-indazole-3-carboxylic acid

  • Cas Number : 129295-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47582.html

Safety Information

MSDS https://synthonix.com/47582.html
Availability details for
Quantity
  • Availability
  • Add to Cart