SY3H3D67A5E9 Synthonix Corporation

6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD02186559
  • Cas Number : 182181-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47583.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47583.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart