SY3H3D67A5EF Synthonix Corporation

6-Chloroquinazolin-4-ol

  • MDL number MFCD00182168
  • Cas Number : 16064-14-5
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-4(3H)-quinazolinone
Inchi Code 1S/C8H5ClN2O/c9-5-1-2-7-6(3-5)8(12)11-4-10-7/h1-4H,(H,10,11,12)
Inchi Key GOBVWEUSCRFCPB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form White to Gray to Yellow to Light-brown Crystal
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47612.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47612.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart