SY3H3D67A5F5 Synthonix Corporation

3-(Cyanomethyl)picolinonitrile

  • Cas Number : 5912-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47626.html

Safety Information

MSDS https://synthonix.com/47626.html
Availability details for
Quantity
  • Availability
  • Add to Cart