SY3H3D67A607 Synthonix Corporation

4-Chloroisophthalic acid

  • Cas Number : 2845-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47820.html

Safety Information

MSDS https://synthonix.com/47820.html
Availability details for
Quantity
  • Availability
  • Add to Cart