SY3H3D67A60B Synthonix Corporation

2-Chloro-1-(3,4-dichlorophenyl)ethanone

  • MDL number MFCD03970382
  • Cas Number : 42981-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47840.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47840.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart