SY3H3D67AC8F Synthonix Corporation

Cyclopropyl(3-methylpyridin-2-yl)methanamine dihydrochloride

  • MDL number MFCD29038796
  • Cas Number : 2061979-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54236.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54236.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart