SY3H3D67AC91 Synthonix Corporation

1-Cbz-Amino-2-methylaminoethane hydrochloride

  • MDL number MFCD11112222
  • Cas Number : 277328-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55521.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55521.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart