SY3H3D67AC93 Synthonix Corporation

3-((4-Chlorobenzyl)oxy)piperidine hydrochloride

  • MDL number MFCD13561019
  • Cas Number : 1220033-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56068.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56068.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart