SY3H3D67AC94 Synthonix Corporation

2-N-Cbz-Butane-1,2-diamine hydrochloride

  • MDL number MFCD11112244
  • Cas Number : 1179361-76-2
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56628.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56628.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart