SY3H3D67AC95 Synthonix Corporation

1-(((4-Chlorophenoxy)carbonyl)oxy)ethyl isobutyrate

  • MDL number MFCD29920546
  • Cas Number : 1622940-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Yellow to Brown Sticky Oil to Semi-Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56940.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56940.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart