SY3H3D67ACA2 Synthonix Corporation

2-(Chloromethyl)-1,3-benzothiazole

  • MDL number MFCD05664944
  • Cas Number : 37859-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Very pale yellow - Reddish yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47967.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47967.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart