SY3H3D67ACD4 Synthonix Corporation

6-Chloro-5-methyl-1H-indazole

  • Cas Number : 1000341-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48334.html

Safety Information

MSDS https://synthonix.com/48334.html
Availability details for
Quantity
  • Availability
  • Add to Cart