SY3H3D67ACDB Synthonix Corporation

(6-Chloro-1H-benzo[d]imidazol-2-yl)methanol

  • MDL number MFCD09971879
  • Cas Number : 6953-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48372.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48372.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart