SY3H3D67ACE4 Synthonix Corporation

2-Chloro-2-phenylacetic acid

  • MDL number MFCD00037753
  • Cas Number : 4755-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Powder with lumps Colour: off-white - yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48589.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48589.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart