SY3H3D67AD06 Synthonix Corporation

5-Chloro-2,3-dihydrobenzofuran

  • Cas Number : 76429-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48731.html

Safety Information

MSDS https://synthonix.com/48731.html
Availability details for
Quantity
  • Availability
  • Add to Cart