SY3H3D67AD0E Synthonix Corporation

2-Chloro-3-cyclopropylpyridine

  • Cas Number : 865664-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48880.html

Safety Information

MSDS https://synthonix.com/48880.html
Availability details for
Quantity
  • Availability
  • Add to Cart