SY3H3D67AD17 Synthonix Corporation

(2-Cyano-4-methylphenyl)boronic acid

  • Cas Number : 1328882-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48898.html

Safety Information

MSDS https://synthonix.com/48898.html
Availability details for
Quantity
  • Availability
  • Add to Cart