SY3H3D67AD25 Synthonix Corporation

1-(4-Chloropyridin-2-yl)propan-1-one

  • MDL number MFCD09863875
  • Cas Number : 896139-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49039.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49039.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart