SY3H3D67AD27 Synthonix Corporation

1-(6-Chloro-5-methylpyridin-3-yl)ethanone

  • Cas Number : 1256791-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49042.html

Safety Information

MSDS https://synthonix.com/49042.html
Availability details for
Quantity
  • Availability
  • Add to Cart