SY3H3D67AD3C Synthonix Corporation

(R)-1-(2-Chloro-6-fluorophenyl)ethanamine

  • Cas Number : 1100575-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49234.html

Safety Information

MSDS https://synthonix.com/49234.html
Availability details for
Quantity
  • Availability
  • Add to Cart